Anti-IL1R2 Mouse Monoclonal Antibody [clone: 1G12]
Lieferant: US Biological
USBI128435EA 733 CHF
USBI128435
Anti-IL1R2 Mouse Monoclonal Antibody [clone: 1G12]
Antikörper
Type: Primary
Antigen: IL1R2 (interleukin 1 receptor, type II)
Clonality: monoclonal
Clone: 1G12
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human