Anti-IL1R2 Rabbit Polyclonal Antibody

Supplier: US Biological
USBI128434EA 750 CHF
USBI128434
Anti-IL1R2 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: IL1R2 (interleukin 1 receptor, type II)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR