Anti-IL36G Mouse Monoclonal Antibody [clone: 8A11]

Supplier: US Biological
USBI128431EA 702 CHF
USBI128431
Anti-IL36G Mouse Monoclonal Antibody [clone: 8A11]
Antibodies
Type: Primary
Antigen: IL36G (interleukin 36, gamma)
Clonality: monoclonal
Clone: 8A11
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR