Anti-IL36B Mouse Monoclonal Antibody [clone: 1E4]

Supplier: US Biological
USBI128430EA 702 CHF
USBI128430
Anti-IL36B Mouse Monoclonal Antibody [clone: 1E4]
Antibodies
Type: Primary
Antigen: IL36B (interleukin 36, beta)
Clonality: monoclonal
Clone: 10000
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR