Anti-IL36B Mouse Polyclonal Antibody

Lieferant: US Biological
USBI128429EA 733 CHF
USBI128429
Anti-IL36B Mouse Polyclonal Antibody
Antikörper
Type: Primary
Antigen: IL36B (interleukin 36, beta)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR