Anti-IL36A Mouse Polyclonal Antibody

Supplier: US Biological
USBI128427EA 702 CHF
USBI128427
Anti-IL36A Mouse Polyclonal Antibody
Antibodies
Type: Primary
Antigen: IL36A (interleukin 36, alpha)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR