Anti-IL1B Rabbit Polyclonal Antibody

Supplier: US Biological
USBI128425EA 750 CHF
USBI128425
Anti-IL1B Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: IL1B (interleukin 1, beta)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR