Anti-IL1A Mouse Monoclonal Antibody [clone: 4B8]

Lieferant: US Biological
USBI128422EA 733 CHF
USBI128422
Anti-IL1A Mouse Monoclonal Antibody [clone: 4B8]
Antikörper
Type: Primary
Antigen: IL1A (interleukin 1, alpha)
Clonality: monoclonal
Clone: 4B8
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR