Anti-IL18RAP Mouse Monoclonal Antibody [clone: 4G4]

Supplier: US Biological
USBI128419EA 702 CHF
USBI128419
Anti-IL18RAP Mouse Monoclonal Antibody [clone: 4G4]
Antibodies
Type: Primary
Antigen: IL18RAP (interleukin 18 receptor accessory protein)
Clonality: monoclonal
Clone: 4G4
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR