Anti-IL18R1 Mouse Monoclonal Antibody [clone: 3C8]

Lieferant: US Biological
USBI128417EA 733 CHF
USBI128417
Anti-IL18R1 Mouse Monoclonal Antibody [clone: 3C8]
Antikörper
Type: Primary
Antigen: IL18R1 (interleukin 18 receptor 1)
Clonality: monoclonal
Clone: 3C8
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR