Anti-IL17F Mouse Monoclonal Antibody [clone: 1E5]

Supplier: US Biological
USBI128412EA 702 CHF
USBI128412
Anti-IL17F Mouse Monoclonal Antibody [clone: 1E5]
Antibodies
Type: Primary
Antigen: IL17F (interleukin 17F)
Clonality: monoclonal
Clone: 100000
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR