Anti-IL17C Mouse Polyclonal Antibody

Supplier: US Biological
USBI128404EA 702 CHF
USBI128404
Anti-IL17C Mouse Polyclonal Antibody
Antibodies
Recommended dilution: Immunochemistry: 3?g/ml

Type: Primary
Antigen: IL17C (interleukin 17C)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR