Anti-IL15RA Mouse Monoclonal Antibody [clone: 1C5]

Lieferant: US Biological
USBI128399EA 733 CHF
USBI128399
Anti-IL15RA Mouse Monoclonal Antibody [clone: 1C5]
Antikörper
Type: Primary
Antigen: IL15RA (interleukin 15 receptor, alpha)
Clonality: monoclonal
Clone: 1C5
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR