Anti-IL15 Mouse Monoclonal Antibody [clone: 2F6]

Supplier: US Biological
USBI128398EA 702 CHF
USBI128398
Anti-IL15 Mouse Monoclonal Antibody [clone: 2F6]
Antibodies
Recommended dilution: Immunochemistry: 1. 5 ?g/ml

Type: Primary
Antigen: IL15 (interleukin 15)
Clonality: monoclonal
Clone: 2F6
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR