Anti-IL13RA2 Rabbit Polyclonal Antibody

Lieferant: US Biological
USBI128394EA 788 CHF
USBI128394
Anti-IL13RA2 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: IL13RA2 (interleukin 13 receptor, alpha 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR