Anti-IL13RA1 Mouse Monoclonal Antibody [clone: 1F5]

Supplier: US Biological
USBI128391EA 702 CHF
USBI128391
Anti-IL13RA1 Mouse Monoclonal Antibody [clone: 1F5]
Antibodies
Type: Primary
Antigen: IL13RA1 (interleukin 13 receptor, alpha 1)
Clonality: monoclonal
Clone: 1F5
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR