Anti-IL13 Mouse Monoclonal Antibody [clone: 3H7]

Lieferant: US Biological
USBI128389EA 733 CHF
USBI128389
Anti-IL13 Mouse Monoclonal Antibody [clone: 3H7]
Antikörper
Recommended dilution: Immunochemistry: 3?g/ml

Type: Primary
Antigen: IL13 (interleukin 13)
Clonality: monoclonal
Clone: 3H7
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR