Drucken

Anti-IL12RB2 Mouse Monoclonal Antibody [clone: 2H6]

Lieferant: US Biological
USBI128388EA 702 CHF
USBI128388
Anti-IL12RB2 Mouse Monoclonal Antibody [clone: 2H6]
Antikörper
Type: Primary
Antigen: IL12RB2 (interleukin 12 receptor, beta 2)
Clonality: monoclonal
Clone: 2H6
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR