Anti-IL12RB1 Rabbit Polyclonal Antibody

Lieferant: US Biological
USBI128385EA 788 CHF
USBI128385
Anti-IL12RB1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: IL12RB1 (interleukin 12 receptor, beta 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR