Anti-IL10 Mouse Monoclonal Antibody [clone: 1C10]

Supplier: US Biological
USBI128379EA 702 CHF
USBI128379
Anti-IL10 Mouse Monoclonal Antibody [clone: 1C10]
Antibodies
Type: Primary
Antigen: IL10 (interleukin 10)
Clonality: monoclonal
Clone: 1C10
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG3 Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR