Anti-IKZF4 Mouse Polyclonal Antibody

Supplier: US Biological
USBI128373EA 702 CHF
USBI128373
Anti-IKZF4 Mouse Polyclonal Antibody
Antibodies
Recommended dilution: Immunofluorescence: 10 ?g/ml

Type: Primary
Antigen: IKZF4 (IKAROS family zinc finger 4 (Eos))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR