Anti-IKZF2 Mouse Polyclonal Antibody

Lieferant: US Biological
USBI128371EA 733 CHF
USBI128371
Anti-IKZF2 Mouse Polyclonal Antibody
Antikörper
Recommended dilution: Immunofluorescence: 10 ?g/ml

Type: Primary
Antigen: IKZF2 (IKAROS family zinc finger 2 (Helios))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR