Anti-NFKBIA Mouse Monoclonal Antibody [clone: 2G4]

Supplier: US Biological
USBI128369EA 702 CHF
USBI128369
Anti-NFKBIA Mouse Monoclonal Antibody [clone: 2G4]
Antibodies
Recommended dilution: Immunofluorescence: 10 ?g/ml; Immunochemistry: 3?g/ml

Type: Primary
Antigen: NFKBIA (nuclear factor of kappa light polypeptide gene enhancer in B-cells inhibitor, alpha)
Clonality: monoclonal
Clone: 2G4
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR