Anti-IKBKE Mouse Monoclonal Antibody [clone: 1F7]

Supplier: US Biological
USBI128367EA 702 CHF
USBI128367
Anti-IKBKE Mouse Monoclonal Antibody [clone: 1F7]
Antibodies
Recommended dilution: Immunochemistry: 3?g/ml

Type: Primary
Antigen: IKBKE (inhibitor of kappa light polypeptide gene enhancer in B-cells, kinase epsilon)
Clonality: monoclonal
Clone: 1F7
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR