Drucken

Anti-IKBKB Rabbit Polyclonal Antibody

Lieferant: US Biological
USBI128365EA 750 CHF
USBI128365
Anti-IKBKB Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: IKBKB (inhibitor of kappa light polypeptide gene enhancer in B-cells, kinase beta)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR