Anti-IKBKB Mouse Polyclonal Antibody

Supplier: US Biological
USBI128364EA 702 CHF
USBI128364
Anti-IKBKB Mouse Polyclonal Antibody
Antibodies
Type: Primary
Antigen: IKBKB (inhibitor of kappa light polypeptide gene enhancer in B-cells, kinase beta)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR