Anti-IKBKAP Mouse Monoclonal Antibody [clone: 6G9]

Supplier: US Biological
USBI128363EA 702 CHF
USBI128363
Anti-IKBKAP Mouse Monoclonal Antibody [clone: 6G9]
Antibodies
Recommended dilution: Immunochemistry: 3?g/ml

Type: Primary
Antigen: IKBKAP (inhibitor of kappa light polypeptide gene enhancer in B-cells, kinase complex-associated protein)
Clonality: monoclonal
Clone: 6G9
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR