Anti-IP6K1 Mouse Polyclonal Antibody

Lieferant: US Biological
USBI128361EA 733 CHF
USBI128361
Anti-IP6K1 Mouse Polyclonal Antibody
Antikörper
Type: Primary
Antigen: IP6K1 (inositol hexakisphosphate kinase 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR