Anti-IHH Mouse Monoclonal Antibody [clone: 2G9]

Supplier: US Biological
USBI128360EA 702 CHF
USBI128360
Anti-IHH Mouse Monoclonal Antibody [clone: 2G9]
Antibodies
Type: Primary
Antigen: IHH (indian hedgehog)
Clonality: monoclonal
Clone: 2G9
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR