Anti-IGSF6 Mouse Monoclonal Antibody [clone: 2A12]

Supplier: US Biological
USBI128359EA 702 CHF
USBI128359
Anti-IGSF6 Mouse Monoclonal Antibody [clone: 2A12]
Antibodies
Type: Primary
Antigen: IGSF6 (immunoglobulin superfamily, member 6)
Clonality: monoclonal
Clone: 2A12
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR