Anti-IGSF21 Mouse Polyclonal Antibody

Supplier: US Biological
USBI128354EA 702 CHF
USBI128354
Anti-IGSF21 Mouse Polyclonal Antibody
Antibodies
Type: Primary
Antigen: IGSF21 (immunoglobin superfamily, member 21)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR