Anti-CD101 Mouse Monoclonal Antibody [clone: 3G12]

Supplier: US Biological
USBI128353EA 691 CHF
USBI128353
Anti-CD101 Mouse Monoclonal Antibody [clone: 3G12]
Antibodies
Type: Primary
Antigen: CD101 (CD101 molecule)
Clonality: monoclonal
Clone: 3G12
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR