Anti-IGSF1 Mouse Monoclonal Antibody [clone: 4C7]

Lieferant: US Biological
USBI128352EA 733 CHF
USBI128352
Anti-IGSF1 Mouse Monoclonal Antibody [clone: 4C7]
Antikörper
Type: Primary
Antigen: IGSF1 (immunoglobulin superfamily, member 1)
Clonality: monoclonal
Clone: 4C7
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR