Anti-RETN Mouse Monoclonal Antibody [clone: RRES 03]

Lieferant: United States Biological
USBI045242EA 526 CHF
USBI045242
Anti-RETN Mouse Monoclonal Antibody [clone: RRES 03]
Antikörper
Type: Primary
Antigen: RETN (resistin)
Clonality: Monoclonal
Clone: RRES 03
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR