Anti-RETN Rat Monoclonal Antibody [clone: MRES 06]

Lieferant: United States Biological
USBI045241EA 540 CHF
USBI045241
Anti-RETN Rat Monoclonal Antibody [clone: MRES 06]
Antikörper
Type: Primary
Antigen: RETN (resistin)
Clonality: Monoclonal
Clone: MRES 06
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a Kappa
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR