Anti-RELM, alpha Mouse Monoclonal Antibody [clone: RREL 803]

Lieferant: United States Biological
USBI045233EA 548 CHF
USBI045233
Anti-RELM, alpha Mouse Monoclonal Antibody [clone: RREL 803]
Antikörper
Type: Primary
Antigen: RELM, alpha
Clonality: Monoclonal
Clone: RREL 803
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR