Anti-RELM-beta Rat Monoclonal Antibody [clone: MRB 46L]

Lieferant: United States Biological
USBI045228EA 540 CHF
USBI045228
Anti-RELM-beta Rat Monoclonal Antibody [clone: MRB 46L]
Antikörper
Type: Primary
Antigen: RELM-beta
Clonality: Monoclonal
Clone: MRB 46L
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a Kappa
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR