Anti-RELM-beta Rabbit Polyclonal Antibody

Lieferant: United States Biological
USBI045222EA 617 CHF
USBI045222
Anti-RELM-beta Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: RELM-beta
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR