Anti-RELM-alpha Mouse Monoclonal Antibody [clone: RREL 804]

Lieferant: United States Biological
USBI045220EA 526 CHF
USBI045220
Anti-RELM-alpha Mouse Monoclonal Antibody [clone: RREL 804]
Antikörper
Type: Primary
Antigen: RELM-alpha
Clonality: Monoclonal
Clone: RREL 804
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR