Anti-RELM-alpha Rabbit Polyclonal Antibody

Lieferant: United States Biological
USBI045215EA 617 CHF
USBI045215
Anti-RELM-alpha Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: RELM-alpha
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR