Anti-Rab6-GTP Human Monoclonal Antibody [clone: AA2]

Lieferant: United States Biological
USBI045191EA 749 CHF
USBI045191
Anti-Rab6-GTP Human Monoclonal Antibody [clone: AA2]
Antikörper
Type: Primary
Antigen: Rab6-GTP
Clonality: Monoclonal
Clone: AA2
Conjugation: Unconjugated
Epitope:
Host: Human
Isotype: IgG2 Lambda
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR