Anti-Progranulin Rat Polyclonal Antibody

Lieferant: United States Biological
USBI045171EA 617 CHF
USBI045171
Anti-Progranulin Rat Polyclonal Antibody
Antikörper
Type: Primary
Antigen: Progranulin
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR