Anti-Polyglutamylation Modification Mouse Monoclonal Antibody (Biotin) [clone: GT335]

Lieferant: United States Biological
USBI045160EA 833 CHF
USBI045160
Anti-Polyglutamylation Modification Mouse Monoclonal Antibody (Biotin) [clone: GT335]
Antikörper
Type: Primary
Antigen: Polyglutamylation Modification
Clonality: Monoclonal
Clone: GT335
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR