Anti-Omentin Rabbit Polyclonal Antibody

Lieferant: United States Biological
USBI045119EA 617 CHF
USBI045119
Anti-Omentin Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: Omentin
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR