Anti-NOTCH2 Rat Monoclonal Antibody [clone: 16F11]

Lieferant: United States Biological
USBI045101EA 665 CHF
USBI045101
Anti-NOTCH2 Rat Monoclonal Antibody [clone: 16F11]
Antikörper
Type: Primary
Antigen: NOTCH2 (notch 2)
Clonality: Monoclonal
Clone: 16F11
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG1 Kappa
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR