Anti-NOTCH1 Rat Monoclonal Antibody [clone: 22E5]
Lieferant: United States Biological
USBI045098EA 665 CHF
USBI045098
Anti-NOTCH1 Rat Monoclonal Antibody [clone: 22E5]
Antikörper
Type: Primary
Antigen: NOTCH1 (notch 1)
Clonality: Monoclonal
Clone: 2200000
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a Kappa
Reactivity:
Order Now