Anti-NOD1 Rabbit Polyclonal Antibody

Lieferant: United States Biological
USBI045096EA 530 CHF
USBI045096
Anti-NOD1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: NOD1 (nucleotide-binding oligomerization domain containing 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR