Anti-NMNAT2 Mouse Monoclonal Antibody [clone: Nady-1]

Lieferant: United States Biological
USBI045091EA 481 CHF
USBI045091
Anti-NMNAT2 Mouse Monoclonal Antibody [clone: Nady-1]
Antikörper
Type: Primary
Antigen: NMNAT2 (nicotinamide nucleotide adenylyltransferase 2)
Clonality: Monoclonal
Clone: Nady-1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR