Anti-NLRP6/NALP6 Mouse Monoclonal Antibody [clone: Clint-1]

Lieferant: United States Biological
USBI045089EA 776 CHF
USBI045089
Anti-NLRP6/NALP6 Mouse Monoclonal Antibody [clone: Clint-1]
Antikörper
Type: Primary
Antigen: NLRP6/NALP6
Clonality: Monoclonal
Clone: Clint-1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR