Anti-NLRP3/NALP3 Mouse Monoclonal Antibody [clone: Cryo-2]

Lieferant: United States Biological
USBI045088EA 851 CHF
USBI045088
Anti-NLRP3/NALP3 Mouse Monoclonal Antibody [clone: Cryo-2]
Antikörper
Type: Primary
Antigen: NLRP3/NALP3
Clonality: Monoclonal
Clone: Cryo-2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR